missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-4-nitrobenzoate, 98%, Thermo Scientific™

Product Code. 11418567
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11418567

Brand: Thermo Scientific Alfa Aesar A18625.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30650-90-9
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.20
MDL Number MFCD00115804
InChI Key OAXJZQMFWBIRKF-UHFFFAOYSA-N
PubChem CID 2796918
IUPAC Name ethyl 3-methyl-4-nitrobenzoate
SMILES CCOC(=O)C1=CC=C(C(C)=C1)[N+]([O-])=O

Specifications

Melting Point 53°C to 55°C
Quantity 10 g
Synonym benzoic acid, 3-methyl-4-nitro-, ethyl ester, 3-methyl-4-nitrobenzoic acid ethyl ester, acmc-209zoi, ethyl 3-methyl-4-nitro-benzoate, benzoic acid,3-methyl-4-nitro-, ethyl ester
Formula Weight 209.2
Percent Purity 98%
Chemical Name or Material Ethyl 3-methyl-4-nitrobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.