missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oxazole-5-carboxylate, 98%

Product Code. 15435707
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435707 5 g 5g
15445707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435707 Supplier Thermo Scientific Alfa Aesar Supplier No. H32924.06
 Restricted Item
Expédition estimée: 08-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 118994-89-1 | C6H7NO3 | 141.126 g/mol

Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.

Solubility
Soluble in water (35 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118994-89-1
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD04114930
InChI Key KRMORCCAHXFIHF-UHFFFAOYSA-N
Synonym ethyl oxazole-5-carboxylate, 5-oxazolecarboxylic acid, ethyl ester, 5-oxazolecarboxylicacid, ethyl ester, oxazole-5-carboxylic acid ethyl ester, ethyloxazole-5-carboxylate, 5-ethoxycarbonyloxazole, acmc-1c1iz, ksc510s9l, ethyl 5-oxazolecarboxylate, 5-ethoxycarbonyl-1,3-oxazole
PubChem CID 10964603
IUPAC Name ethyl 1,3-oxazole-5-carboxylate
SMILES CCOC(=O)C1=CN=CO1

Specifications

Boiling Point 60°C (2.25 mmHg)
Refractive Index 1.469
Quantity 5 g
Beilstein 83084416
Solubility Information Soluble in water (35g/L at 25°C).
Formula Weight 141.13
Percent Purity 98%
Chemical Name or Material Ethyl oxazole-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.