Learn More
Thermo Scientific™ Eutech™ Sensor for ECTestr11
Product Code. 10475982
Change view
Click to view available options
For Use With (Equipment):
ECTestr11
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10475982 ECTestr11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10475982 Supplier Thermo Scientific™ Supplier No. 01X229714
  • ONLINE EXCLUSIVE PRICE
    84.90€
    77.30€ / 1 pièces
    Économisez 7.60€
    8% de réduction
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
Eutech™ offers meters that allow the direct reading of TDS values. Eutech™ Sensor for ECTestr11 is a replacement sensor for waterproof ECTestr11, Cup Type. Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
General applications:
  • water and wastewater treatment
  • boiler blow-down
  • electroplating rinse tanks
  • drinking water
  • hydroponics
  • printing industry
  • aquaculture, aquariums, fish farms
  • swimming pools and spas
TRUSTED_SUSTAINABILITY
Specifications
Type Sensor
For Use With (Equipment) ECTestr11