Learn More
Neogen Extendable Pole XSL008
Product Code. 17288520
Encompass
Change view
Click to view available options
Length (English):
8 ft.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
17288520 8 ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17288520 Supplier Neogen Supplier No. 7000030316
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Extendable aluminum pole with Neogen Sponge-Stick holder.
  • Telescoping 8 ft 5 in. (2.56 m) pole.
  • Includes Neogen Sponge-Stick holder.
Specifications
Product Type Extendable Sponge Pole
Length (English) 8 ft.
Packaging Quantity 1 Ea.