Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Heavy Solvents
Product Code. 10661702
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10661702 500 mL Each
10654582 100 mL Each
10151471 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10661702 Supplier Lenz Laborglasintrumente Supplier No. 05433158
  • ONLINE EXCLUSIVE PRICE
    449.00€
    296.39€ / 1 pièces
    Économisez 152.61€
    33% de réduction
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For the extraction of liquids with specific heavy solvents
  • With insert filter (porosity: 0)
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 500 mL
Material Fritte P0
Standard Taper Joint NS 60/46