Learn More
Kartell Eye Wash Station (German Version)
Product Code. 18037821
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
18037821 Green Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18037821 Supplier Kartell Supplier No. 0238700
  • ONLINE EXCLUSIVE PRICE
    270.00€
    251.92€ / 10 pièces
    Économisez 18.08€
    6% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Emergency eye wash station moulded in strong high-impact PS. The directions are clearly printed on the support rack and the bottle - to be bought separately - can be filled with distilled water or other medicated liquid for ophthalmic use.
  • Emergency eye wash station moulded in strong high-impact PS
  • The directions are clearly printed on the support rack and the bottle can be filled with distilled water or other medicated liquid for ophthalmic use
  • The head has an air-vent valve to assure air refill of free space in the bottle thus preventing draw back of contaminated wash into the bottle
  • A perforated spray plate in the cup centre breaks up the stream to give the eye a gentle wash
  • A drain tube releases waste
Specifications
Color Green
Material Polystyrene
Mounting Wall
Product Type Eye Wash Station
Sterility Non-sterile