Learn More
Gardco™ EZ/3 Series Viscosity Cup #3 with Certificate
Product Code. 11775359 Shop All Cole-Parmer Products
Change view
Click to view available options
Viscosity Range:
64 to 596 cSt
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Unit Size
11775359 64 to 596 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11775359 Supplier Gardco™ Supplier No. 0870022
  • ONLINE EXCLUSIVE PRICE
    612.00€
    575.07€ / 1 pièces
    Économisez 36.93€
    6% de réduction
Expédition estimée: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Gardco EZ3/C series viscosity cup #3 with certificate is an easy and economical way to measure viscosity.
  • Supplied with conversion table to determine viscosity in centistokes (cSt)
  • Meets dimensions as specified in ASTM D 4212 standards
  • NIST Traceable certificate
Specifications
Viscosity Range 64 to 596 cSt
Description Gardco EZ3/C Series Viscosity Cup #3
Certifications/Compliance Cup dimensions as specified in ASTM D 4212 standards; NIST-traceable models conform to MIL STD 5662A, ANSI/NCSL 2540-1; Traceable to NIST-standard oils; NIST-traceable NIST Certificate
Volume (Metric) 46.5 mL