Learn More
Julabo™ F Series Recirculating Cooling Baths with Air Compressor
Product Code. 11994858
Change view
Click to view available options
Capacity (Metric):
2.6 L
7.5 L
9.5 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11994858 2.6 L Each
15194342 7.5 L Each
15214373 9.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: This product is under eco-contribution tax.
Product Code. 11994858 Supplier Julabo™ Supplier No. 9620025
  • ONLINE EXCLUSIVE PRICE
    3626.01€
    2665.00€ / 1 pièces
    Économisez 961.01€
    26% de réduction
View all promotions
 Restricted Item
En stock
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: This product is under eco-contribution tax.
Require very little laboratory space and have very low procurement costs. . Julabo™ F Series Recirculating Cooling Baths with Air Compressor are a great way to replace costly systems based on running tap water and are ideal for basic cooling tasks.
  • Environmentally friendly operation with low energy consumption
  • Compact design
  • Splashproof membrane keypad with LED temperature display
  • Straightforward filling and draining
  • Filling level indicator
  • May be used with water, water/glycol, Julabo Thermal G
Specifications
Capacity (Metric) 2.6 L
Certifications/Compliance UL Listed and NRTL marked
Cooling Capacity 0.09kW at -5°C, 0.18kW at 0°C, 0.21kW at 5°C, 0.22kW at 10°C, 0.25kW at 20°C (Me
Flow Rate 15 L/min.
Temperature Range -10°C to +40°C
Max. Pressure 50.7 psi
Model F250
Refrigerant R134A
Temperature Accuracy ±0.5°C
Temperature Stability ±0.5°C
Length (Metric) 40 cm
Width (Metric) 24 cm
Height (Metric) 52 cm
Shipping Weight (Metric) 27.66 kg
Weight (Metric) 27 kg
Electrical Requirements 115/230 V 60 Hz 6 A
Voltage 115/230 V
Frequency 60 Hz