Learn More
Moldex™ Face Masks
Product Code. 12982415
Change view
Click to view available options
Size:
Medium/Large
Small
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12982415 Small Pack of 10
12992415 Medium/Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12982415 Supplier Moldex™ Supplier No. 315501
  • ONLINE EXCLUSIVE PRICE
    72.35€
    48.40€ / 10 pièces
    Économisez 23.95€
    33% de réduction
Expédition estimée: 30-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The filtering surface is almost three times larger than a standard filter product
The uniquely developed Series 3000 Air from Moldex offers extremely low breathing resistance and therefore increased performance of both mask and wearer. The filtering surface is almost three times larger than a standard filter product, allowing you to breathe more easily.
Specifications
Color White
Product Type Face Mask
Size Small
Quantity 10/Pk.