Learn More
Kartell™ Sterile Faeces Container
Product Code. 10244331
Change view
Click to view available options
Volume (Metric):
60 mL
Unit Size:
Pack of 400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10244331 60 mL Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10244331 Supplier Kartell™ Supplier No. 0269500
  • ONLINE EXCLUSIVE PRICE
    256.95€
    153.26€ / 400 pièces
    Économisez 103.69€
    40% de réduction
 Restricted Item
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Easy means of collecting and transporting faeces samples. Kartell™ Sterile Faeces Container includes a spoon to assist in handling the sample.
  • Particularly designed for collection and delivery of faeces
  • Made of transparent PS with stopper of PP
  • Incorporates a small spoon to facilitate sample handling
  • Sterile
  • CE Marked and IVD Certified
Specifications
Certifications/Compliance CE/IVD
Diameter (Metric) 41 mm
Includes PP Stopper and Spoon
Material Polystrene
Packaging Sterile
Volume (Metric) 60 mL
Height (Metric) 55 mm