Learn More
Fenchlorphos, MedChemExpress
Product Code. 16448606
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448606 Supplier Medchem Express Supplier No. HYB1093
 Restricted Item
Expédition estimée: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Fenchlorphos, an organophosphate, is an insecticide. Fenchlorphos is an inhibitor of the enzyme acetylcholinesterase (AChE). Fenchlorphos is able to cause mitochondrial dysfunction.
Fenchlorphos, an organophosphate, is an insecticide. Fenchlorphos is an inhibitor of the enzyme acetylcholinesterase (AChE). Fenchlorphos is able to cause mitochondrial dysfunction.
Specifications
Chemical Name or Material Fenchlorphos
CAS 299-84-3
Color White
Purity Grade Notes Research
Molecular Formula C8H8Cl3O3PS
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (310.99 mM; Need ultrasonic)
SMILES S=P(OC)(OC)OC1=CC(Cl)=C(Cl)C=C1Cl
Molecular Weight (g/mol) 321.55
Formula Weight 321.55
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation