Learn More
Thermo Scientific™ FH 40 G Multi-Purpose Digital Survey Meter
Product Code. 17952282 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Digital Survey Meter
Display Unit
Measurement Range:
0.1 μSv/h to 0.99Sv/h, 36keV tp 1.3MeV
N/A
Up to 1 Sv/h
Up to 100 mSv/h
Units:
Selectable units: Sv/h, R/h, Gy/h
Sv/h
Includes:
Extra earphone connector
No extra earphone connector
No internal counter tube
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Measurement Range Units Includes Unit Size
17952282 Digital Survey Meter Up to 100 mSv/h Sv/h Extra earphone connector Each
18890374 Digital Survey Meter 0.1 μSv/h to 0.99Sv/h, 36keV tp 1.3MeV Selectable units: Sv/h, R/h, Gy/h No extra earphone connector Each
17857566 Digital Survey Meter Up to 1 Sv/h Sv/h Extra earphone connector Each
13417416 Digital Survey Meter Up to 100 mSv/h Sv/h No extra earphone connector Each
18850422 Display Unit N/A Sv/h No internal counter tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
FH 40 G and FH 40 G-L series meters (4254002, 425400201, 425400250, 425400251, 4254004, 425400401, 425400450, 425400451) are not for sale in the UK
Regulatory: This product is under eco-contribution tax.
Product Code. 17952282 Supplier Thermo Scientific™ Supplier No. 425400850
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
FH 40 G and FH 40 G-L series meters (4254002, 425400201, 425400250, 425400251, 4254004, 425400401, 425400450, 425400451) are not for sale in the UK
Regulatory: This product is under eco-contribution tax.
Use one convenient, versatile instrument for multiple radiation detection applications including personal radiation protection and beta, x-ray and gamma radiation detection.
Serve multiple radiation detection applications with one versatile instrument. The Thermo Scientific™ FH 40 G is a wide range digital Geiger counter suitable for nearly all measurement tasks arising in radiation protection through optional plug and play probes available for neutron measurement, alpha and beta contamination readings and even to detect artificial gamma radiation in variable natural radiation fields.
The FH 40 G is a versatile multipurpose meter and area monitor that can be used with external detectors for portable and remote area monitoring applications.
  • Rugged, waterproof and reliable with very simple operation.
  • Gamma meter with internal proportional counter tube.
  • Integrated gamma dose rate measurements.
  • Interface for one optional, external plug and play probe.
  • Alarm thresholds for the internal detector and external probes of the FH 40 G are monitored simultaneously.
  • Upon connection of an external plug and play probe, the probe parameters are automatically read into the FH 40 G and the correct parameters are set.
  • High voltage generation inside the plug and play probe makes the system calibration independent of cable length.
  • Separate alarms can be set by the user for both the internal and external detectors.
  • Operational internal proportional detector and connected external detector.
  • Serial infrared interface.
  • Type tested by PTB (German version).
  • Type tested for use by German fire brigades.
  • Simultaneous operation of internal and external detectors, both dose rate and remote dose rate readings.
  • Configuration and data read out via PC.
  • Waterproof (IP 67) and shock resistant.
  • ADF mode (Advanced Digital Filter).
  • Clear, backlit LCD display.
  • Datalogging capability.
  • Internal diagnostics ensure proper functioning of the detector(s) and the electronics.
  • Designed to record up to 256 data points containing measurement number, date, time and dose rate at the internal detector and external detector, status, and optional barcode information.
  • Wide range of plug and play external probes for monitoring and detection.
  • All FH 40 G versions and external probes containing the "-10" suffix are designed to meet the energy response behavior of the new SI-units ambient dose equivalent and ambient dose equivalent rate according to ICRU 39.
Short summary of available plug and play probes:
  • Gamma dose rate measurement: FHT 192 (ionization chamber), FHZ 632 (proportional counter), FHZ 612 (wide range), FHZ 302/312 (under water nuclear power).
  • Gamma search probes: FHZ 502, 503, 512 (NaJ/BGO).
  • Contamination: FHZ 742 (Scintillator), FHZ 732 (Pancake/proportional counter), FHZ 382 (Scintiallator + Alpha/Beta discriminator via LED indication if alphas are present).
  • Aritificial radiaiton discrimination: NBR probe FHZ 672.
  • Preamplifiers for various classic probes.
  • Wide range of accessories like cables, telepoles, application specific kits, etc.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets IP 67 requirement for waterproofing; shock-resistence / NRPB approved (UK); ICRU 39 for SI-units
Description FH 40 GL-10 Radiameter energy compensated for ambient dose equivalent H*(10) according to ICRP60/ICRU39
Calibration Export version with selectable calibration factor
Energy Range FH 40 G: 36 keV to 1.3 MeV; FH 40 G-L: 36 keV to 1.3 MeV; FH 40 G-10: 30 keV to 4.4 MeV; FH 40 GL-omega: 36 keV to 1.3 MeV
Product Type Digital Survey Meter
Measurement Range Up to 100 mSv/h
Height (English) 1.7 in.
Measured Quantity FH 40 G, FH 40 G-L, FH 40 GL-omega: Response (Roentgen) - R; FH 40 G-10: Ambient dose equivalent rate (H™(10)) - Sv
Width (Metric) 7.4 cm
Weight (Metric) 0.41 kg
Radiation Type Dependent on probe used
Units Sv/h
Depth (Metric) 195 mm
Width (English) 2.9 in.
Weight (English) 0.9 lb.
Includes Extra earphone connector
Depth (English) 7.7 in.
Height (Metric) 4.2 cm
Dose Range FH 40 G: 10 µR - 1000 R; FH 40 G-L, FH 40 GL-omega: 10 µR - 100 R; FH 40 G-10: 100 nSv - 10 Sv
Sensitivity > 30 keV