Learn More
Thermo Scientific™ Fiberlite F14-14 x 50cy for LYNX 4000 and 6000 Superspeed Centrifuges
Product Code. 13431369
Change view
Click to view available options
Capacity:
14 x 50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
13431369 14 x 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13431369 Supplier Thermo Scientific™ Supplier No. 096145075
  • ONLINE EXCLUSIVE PRICE
    10690.00€
    8489.47€ / 1 pièces
    Économisez 2200.53€
    20% de réduction
View all promotions
 Restricted Item
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Shorten set-up time and experience peace of mind that the rotor is secure with the Thermo Scientific™ Fiberlite™ F14-14 x 50cy Fixed-Angle Rotor.
Shorten set-up time and experience peace of mind that the rotor is secure with the Thermo Scientific™ Fiberlite™ F14-14 x 50cy Fixed-Angle Rotor. Ideal for pelleting cells, tissue homogenates or subcellular separations at high speeds, this carbon fiber rotor supports 50mL conical tubes, with adapters for 15mL or other tubes.
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Makes lifting and carrying of rotors easier and safer, further enhanced with the lightweight design
Improved Ergonomics and Performance:
  • Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal.
  • Corrosion and fatigue-resistance secures the rotor's structural integrity for unequalled durability.
  • Rotor is backed by a 15-year warranty.
TRUSTED_SUSTAINABILITY
Specifications
Description Fiberlite F14-14x50cy Fixed Angle Rotor
Material Carbon Fiber Composite
Type Fixed Angle Rotor
Capacity 14 x 50 mL
Max. RCF 33,746 x g
Max. Speed 14,000 rpm
Radius (Metric) 154 mm
Radius (Metric) Min. 83 mm
K Factor 798
Max. Tube Length 115 mm
Max. Tube Diameter 29 mm
Net Weight 17.5 lb. (8.0 kg)
For Use With (Equipment) Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle 34°
Biocontainment Yes