Learn More
Gibco™ Fibronectin Human Protein, Plasma
Product Code. 11564466 Shop All Gibco Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11564466 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11564466 Supplier Gibco™ Supplier No. 33016015
  • ONLINE EXCLUSIVE PRICE
    1842.00€
    1639.65€ / 5mg
    Économisez 202.35€
    10% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Fibronectin Human Protein, Plasma is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules.
Fibronectin is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules. Human Plasma Fibronectin is isolated from human plasma by affinity chromtography. Fibronectin has a molecular weight of 440,000 and is made up of two 220,000 dalton polypeptides. Human Fibronectin is filtered (0,2UI) and supplied lyophilized.
Performance and Quality Testing
The purity is determined by SDS-PAGE. Bovine and human fibronectins are assayed for their ability to support cell attachment and spreading, and screened to confirm absence of mycoplasma or bacterial contamination.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cell Culture
Quantity 5 mg
Packaging Glass Bottle
Gene Alias Fibronectin Human, Plasma
Species Human
Recombinant Native
Sterility Sterile
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Fibronectin
Research Category Extracellular Matrix & Cell Adhesion, Stem Cell Research
Color White
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures.