Learn More
Fisherbrand™ Computer Data Acquisition System for Digital Barometer
Product Code. 15567603 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand Traceable Digital Barometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15567603 Fisherbrand Traceable Digital Barometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15567603
 Restricted Item
Expédition estimée: 08-02-2027
Add to basket
Add to basket
Keep track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more. Computer Data Acquisition System for Digital Barometer is for use with Fisherbrand™ Traceable™ Digital Barometer.
  • Allows up to eight unique Traceable™ meters to simultaneously capture data and send it to directly to email to any smartphone or computer
  • Permanently capture readings; keep track of results
  • Readings are saved to a file that can be viewed, printed as-is, or merged in a report or spreadsheet format
  • Data continues to be collected in background even when user works on other software programs
  • 24/7 audible/visual alarm reminds user to check on data
Specifications
Type Data Acquisition Software
Display Type No Display
Software Type Computer Data Acquisition System
Includes CD, 6 ft. Cable
For Use With (Application) Easily keeps track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more
For Use With (Equipment) Fisherbrand Traceable Digital Barometer
Length (English) Cable 6 ft.