Learn More
Fisherbrand™ Hands-Free Fold-Up Magnifiers
Product Code. 11794396 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Magnification:
10x
2.5x
3.5x
3x
5x
7x
Diameter (Englis) Lens:
0.5 in.
0.87 in.
1.25 in.
2 in.
3 in.
3.5 in.
4 in.
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Diameter (Englis) Lens Unit Size
11794396 7x 0.87 in. Each
11744396 2.5x 4 in. Each
11754396 2.5x 3.5 in. Each
11764396 3x 3 in. Each
11704406 10x 0.5 in. Each
11784396 5x 1.25 in. Each
11774396 3.5x 2 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11794396
Quantity Price
1 36.87€ / 1 pièces
10 32.48€ / 1 pièces
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Includes
Batteries, with lighted models.
Allow two-handed examination of objects while yielding a sharp, crisp and distinct image
Fisher Scientific™ Hands-Free Fold-Up Magnifiers all fold twice for compact storage. Allow two-handed examination of objects while yielding a sharp, crisp and distinct image.
  • Bottom panel features white grid markings in both inches and millimeters to assist in measuring
  • All-glass optics ensure perfect focus with zero distortion
  • Seven sizes are available, including three featuring brilliant battery-powered illumination
Specifications
For Use With (Application) For use in quality control, labs, food, electronics, manufacturing and universities
Diameter (Englis) Lens 0.87 in.
Diameter (Metric) Lens 2.21 cm
Magnification 7x
Material Glass
Product Type Magnifier (Hands-free, Fold-up)