Learn More
Fisherbrand™ Clear Plastic Wrap
Product Code. 11785873 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
1 Roll
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11785873 1 Roll Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11785873
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
En stock
Add to basket
Edge
Add to basket
Designed to maintain sample purity.
Designed specifically for the laboratory
  • Made of 100% ultraclean polyethylene—ensures sample purity and eliminates contamination
  • High-cling properties make it ideal for wrapping of beakers, flasks, or Petri dishes with an extraordinarily tight seal that stays in place
  • Low-tangle properties ensure wrap won't ball up when being handled
  • Carton has convenient metal tear-off bar
Specifications
Applications Sealing, Weighing, Transferring Samples
Length (English) 100 ft.
Color Clear
Product Type Wrapping Film
Width (English) 12 in.
Width (Metric) 30cm
Length (Metric) 30.4 m
Material Polyethylene
Quantity 1 Roll