Learn More
Bochem™ Flat Bottom Bowl
Product Code. 10682971
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10682971 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10682971 Supplier Bochem™ Supplier No. 8671
Expédition estimée: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Bochem™ Flat Bottom Bowl is constructed of Stainless Steel for long lasting durability.
  • Stainless Steel
  • Flat bottom
Specifications
Material Stainless Steel (18/10)
Capacity (Metric) 250 mL
Height (Metric) 50 mm
Diameter (Metric) 100 mm
Shape Flat Bottom