Learn More
TPP™ Flow Cytometer Caps
Product Code. 11341724
Change view
Click to view available options
Product Type:
13.5 mm diameter cap
Unit Size:
Pack of 150
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11341724 13.5 mm diameter cap Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11341724 Supplier TPP™ Supplier No. 87008
  • ONLINE EXCLUSIVE PRICE
    72.45€
    37.00€ / 150 pièces
    Économisez 35.45€
    48% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Manufactured with polyethylene for strength. TPP™Flow Cytometers Caps secure PVC packed cell volume tubes for centrifugation
Manufactured with polyethylene for strength. TPP™Flow Cytometers Caps secure PVC packed cell volume tubes for centrifugation
  • For TPP Flow Cytometers
Specifications
Product Type 13.5 mm diameter cap