Learn More
Hellma Flow-Through Cell, 3-in-1, 177-QS
Product Code. 15859824
Change view
Click to view available options
Capacity (Metric):
110 μL
550 μL
Path Length:
10 x 1 mm
10 x 5 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Path Length Unit Size
15859824 550 μL 10 x 5 mm Each
15786962 110 μL 10 x 1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15859824 Supplier Hellma Supplier No. 1767601540
  • ONLINE EXCLUSIVE PRICE
    965.00€
    890.00€ / 1 pièces
    Économisez 75.00€
    7% de réduction
Expédition estimée: 17-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Highly pure Suprasil™ quartz glass flow-through cuvette for absorbance, transmission, and fluorescence measurements and designed with 2 FEP tubing screw connections.
  • High-purity synthetic fused silica quartz construction
  • UV/VIS range
  • Integrated screw connections for FEP tubing
Specifications
Capacity (Metric) 550 μL
Material Suprasil Quartz
Width (Metric) Interior 5 mm
Includes Two Screw Connections, FEB Tubing
Quantity 1/Pk
Path Length 10 x 5 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 35 mm
Width (Metric) 12.5 mm