Learn More
WTW™ Flow-through Vessel for WTW™ TetraCon™ 325
Product Code. 10737831
Change view
Click to view available options
Capacity (Metric):
13 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10737831 13 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10737831 Supplier WTW™ Supplier No. 203730
  • ONLINE EXCLUSIVE PRICE
    702.00€
    610.00€ / 1 pièces
    Économisez 92.00€
    13% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine the conductivity of solutions in closed circuits.The overpressure in the system must not exceed 0.5 bar.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 13 mL
Color Transparent
Description Flow-through vessel
For Use With (Equipment) CellOx™ 325 or TetraCon™ 325 Conductivity Cell
For Use With (Application) For ultrapure water measuring
Material PVC