Learn More
3M™ Flowstream™ Air Regulator System
Product Code. 11771353 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11771353 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11771353 Supplier 3M™ Supplier No. 7100005806
  • ONLINE EXCLUSIVE PRICE
    622.00€
    423.06€ / 1 pièces
    Économisez 198.94€
    31% de réduction
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Flowstream Regulator
For Use With (Equipment) working pressure of 3-8 bar
Includes Integrated warning whistle, integrated activated charcoal filter, belt swivel mechanism, auxillary connection for air tools, integrated silencer (<65dBA)
Quantity 1 Ea.