Learn More
Fmoc-3-(2-naphthyl)-D-alanine, TRC
Product Code. 30178939
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30178939 5 g 5g
30179220 1 g 1g
30179444 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30178939 Supplier Toronto Research Chemicals Supplier No. TRCF6250505G
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 138774-94-4
Molecular Formula C28 H23 N O4
Molecular Weight (g/mol) 437.49
Synonym (R)-α-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-2-naphthalenepropanoic Acid
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
SMILES OC(=O)[C@@H](Cc1ccc2ccccc2c1)NC(=O)OCC3c4ccccc4c5ccccc35
Specifications
InChI Formula InChI=1S/C28H23NO4/c30-27(31)26(16-18-13-14-19-7-1-2-8-20(19)15-18)29-28(32)33-17-25-23-11-5-3-9-21(23)22-10-4-6-12-24(22)25/h1-15,25-26H,16-17H2,(H,29,32)(H,30,31)/t26-/m1/s1
Quantity 5 g
Formula Weight 437.163
Shipping Condition Room Temperature
Chemical Name or Material Fmoc-3-(2-naphthyl)-D-alanine