Learn More
Motic™ Widefield Eyepiece with Diopter Adjustment for AE Microscope
Product Code. 11877993
Click to view available options
Magnification Power:
15X
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 11877993
Brand: Motic™ 1101001402061
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Focusable eyepiece increases magnification. Motic™ Widefield Eyepiece with Diopter Adjustment is for use with an inverted AE microscope.
  • For use with AE Series Microscopes
  • Magnification/Field Number: 12.5X/18mm or 15X/16mm
  • Includes Diopter Adjustment
Order Info
dimProductType
Specifications
Type Focusable Widefield Eyepiece
Field of View 16 mm
Magnification Power 15X
Quantity 1 Ea.