Learn More
Testo™ Food Penetration Probe
Product Code. 10760433
Change view
Click to view available options
Description:
Food Penetration Probe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10760433 Food Penetration Probe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 10760433 Supplier Testo™ Supplier No. 0603 2492
  • ONLINE EXCLUSIVE PRICE
    155.93€
    117.00€ / 1 pièces
    Économisez 38.93€
    24% de réduction
Expédition estimée: 04-12-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Specifications
Description Food Penetration Probe
For Use With (Equipment) testo 926, testo 735
Type Thermocouple type T
For Use With (Application) Measuring the core temperature of food
Includes Fixed 1.5 m cable
Length (Metric) 114.3 mm