Learn More
3M™ Forestry Headgear Combination
Product Code. 15739867 Shop All 3M Products
Change view
Click to view available options
Color: Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15739867 Color: Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15739867 Supplier 3M™ Supplier No. 7000103790
  • ONLINE EXCLUSIVE PRICE
    1025.00€
    782.26€ / 12 pièces
    Économisez 242.74€
    23% de réduction
Expédition estimée: 05-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
3M™ Forestry Headgear Combination provides a versatile and comfortable solution for wearers who require both face and hearing protection.
  • A practical, convenient solution for face and hearing protection where no head protection is needed
  • Front cover provides ventilation for increased comfort and also designed as a cable holder for combination with communication products
  • Mesh Face Shield: + 5°C (Stainless Steel) is approved for High Speed Particles 45 m/s (F) according to EN1731
  • To increase eye protection, G500 can incorporate the 3M™ V6 integrated safety glasses range, that are available in clear, gray and yellow lenses
  • Headgear can be used both with and without earmuffs
  • Headband is slotted for the attachment of earmuffs
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Forestry
Product Type Headgear Combination