Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Product Code. 15793235
This item is not returnable. View return policy
Product Code. 15793235 Supplier Eppendorf™ Supplier No. 6001012510
  • ONLINE EXCLUSIVE PRICE
    199.00€
    181.00€ / 1 pièces
    Économisez 18.00€
    9% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), 5 in/127 mm, drawer, stainless steel- Rack for 127 mm/5 in boxes (15/50 mL conical tubes)- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 580 x 140 x 414 (22.8 x 5.5 x 16.3 in)- Max. box number per rack: 12 (3 x 4)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 108 boxes), Innova U535 (max. 144 boxes), Innova U725 and U725-G (max. 216 boxes), CryoCube F740 series (3-compartment version; max. 216 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack