Learn More
Vileda Professional™ CE Sieve Support Plate
Product Code. 17892486
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17892486 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17892486 Supplier Vileda Professional™ Supplier No. 143776
  • ONLINE EXCLUSIVE PRICE
    212.00€
    183.21€ / 1 pièces
    Économisez 28.79€
    13% de réduction
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Trolley's the meet the highest cleanroom standards! The CE trolley range has been specifically designed and developed for use in controlled environments where maximum contamination control is needed.
  • For use with the CE Dip-N-Go Sieve for the CE Pre-Prepared Trolley
  • Stainless Steel
  • Autoclavable
Specifications
Product Type CE Sieve Support Plate
Autoclavable Autoclavable
Color Gray
Dimensions 55 x 17 x 8 cm (22 x 7 x 3 in.)
For Use With (Equipment) CE Pre-Prepared Trolley with the CE Dip-N-Go Sieve