Learn More
IKA™ Fritte Set
Product Code. 17583626 Shop All IKA Products
Change view
Click to view available options
Size:
1 μm
5 μm
10 μm
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17583626 10 μm Pack of 2
17563626 1 μm Pack of 2
17573626 5 μm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17583626 Supplier IKA™ Supplier No. 0020102634
Expédition estimée: 02-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
This Fritte Set contains two glass frits for the Pro-Divide reaction cell.
  • This set contains two glass frits for the Pro-Divide reaction cell.
  • The size of each pore falls between these ranges:
    • 1 μm set: 1 and 1.6 μm
    • 5 μm set: 4 and 5.5 μm
    • 10 μm set: 10 and 16 μm
TRUSTED_SUSTAINABILITY
Specifications
Type Fritte Set
Size 10 μm
For Use With (Equipment) Pro-Divide Reaction Cell