Learn More
Cole-Parmer™ FSB-200 Series Compact Fluidized Sand Baths
Product Code. 16319501
Change view
Click to view available options
Temperature Range:
50°C to 600°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Unit Size
16319501 50°C to 600°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 16319501 Supplier Cole-Parmer™ Supplier No. 0118950
  • ONLINE EXCLUSIVE PRICE
    8160.00€
    7248.20€ / 1 pièces
    Économisez 911.80€
    11% de réduction
View all promotions
Expédition estimée: 09-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Fast heat-ups and accurate temperature control to 600°C.
  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation
  • Safer alternative to high temperature oil and salt baths
  • Requires controller (01184-50) in order to be calibrated
Use these fluidized sand baths for your heat treating processes, temperature sensor calibrations, and electrical component testing. Baths use aluminum oxide fluidized by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, nonflammable aluminum oxide is safer to use than oil and will not corrode your sample vessels or produce toxic fumes. Vessels of all shapes can be easily immersed and safely removed from the bath, without the hazard and mess of hot oils. Make temperature adjustments via a control knob and reference dial. Baths require a dry air flow of approximately 2 cubic feet per minute (57 L/min) at about 3 psi pressure. Order an air pump separately. Order temperature controller (01184-50 or 01184-55) to accurately control the temperature of your sand bath. Baths feature a thermowell to hold a thermocouple thermometer. Note: not recommended for shape setting in laboratory research device manufacturing or for heating of reactors.
Specifications
Description Compact Fluidized Sand Bath
Temperature Range 50°C to 600°C
Type Sand Bath
Heat Up Time 105 Minutes
Material Stainless Steel
Height (English) 28 in.
Height (Metric) 69.5 cm
Depth (English) Interior 13.75 in.
Voltage 230 VAC
Frequency 50/60 Hz