Learn More
Fungilab™ Stainless Steel Spindles
Product Code. 11896631
Change view
Click to view available options
Includes:
L2 Spindle
TR8, TR9, TR10, TR11 Spindle
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
11896631 TR8, TR9, TR10, TR11 Spindle Each
12827142 L2 Spindle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11896631 Supplier Fungilab™ Supplier No. V12700N
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Accurately measure viscosities with these stainless steel spindles. Stainless Steel Spindles come in a variety of volumes and viscosity ranges.
Spindles for use with Small Sample Adapters (Cat. Nos. 06-815-032, -031).
  • This system is used for measurements in accordance with the ASTM-ISO 2555 standard
  • Standard spindles (included in standard delivery): R and H spindles, L spindles
  • Special spindles (additionally ordered with small sample adapters APM and APM/B): TL and TR spindles
  • Spindle set L: 20 to 6,000,000 cP (for ADV and PRO models); 20 to 2,000,000 cP (for ONE model)
  • Spindle set R: 100 to 40,000,000cP
  • Spindles set R for H models: 200 to 106,000,000cP
Specifications
Product Type Spindle Set
Viscosity Range R: 40 to 400.000/H: 200-3.200.000cP
For Use With (Equipment) Small Sample Adapter
Description Set of special spindles for APM nad Thermosphere (TR8, TR9,TR10,TR11)
Certifications/Compliance ASTM, ISO 2555
Includes TR8, TR9, TR10, TR11 Spindle
Material Stainless Steel
For Use With (Application) For relative viscosity measurement