Learn More
Usbeck Laborgeraete™ Galvanized Steel Support Rod for Stand Base
Product Code. 10211831
Change view
Click to view available options
Length (Metric):
550 mm
600 mm
750 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10211831 600 mm Each
10232911 750 mm Each
10638752 550 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10211831 Supplier Usbeck Laborgeraete™ Supplier No. 2102
  • ONLINE EXCLUSIVE PRICE
    8.85€
    8.00€ / 1 pièces
    Économisez 0.85€
    9% de réduction
 Restricted Item
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Support your M10 laboratory stand bases with Usbeck Laborgeraete™ Galvanized Steel Support Rod for Stand Base. The rod has an internal diameter of 12mm.
  • Steel, zinc plated
  • Threaded M 10
  • Ø 12 mm
Specifications
Material Galvanized Steel
Length (Metric) 600 mm
Diameter (Metric) Inner 12 mm
Product Type Support Rod
For Use With (Equipment) M10 stand base
Quantity 1 Ea.