Learn More
BW Technologies™ Gas Detectors
Product Code. 11968995
Click to view available options
Components Measured:
Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide, VOC, Carbon Dioxide Gas and Fuel
Oxygen, LEL
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11968995
Brand: BW Technologies™ QTXW00RYE00
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Alarm Types Audible
Battery Life 20 hr.
Battery Type Lithium, Rechargeable
Certifications/Compliance Ex II 1G Ga Ex ia IIC T4
Depth (Metric) 47 mm
Components Measured Oxygen, LEL
Type Gas Alert Dectector
Height (Metric) 130 mm
Length (Metric) 81 mm
Measurement Range 0 to 100%
Resolution 1 to 0.1 %
Warranty 24Months
Weight (Metric) 316g