Learn More
Thermo Scientific™ GC Syringe for CTC
Product Code. 15294563 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15294563 2.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15294563 Supplier Thermo Scientific™ Supplier No. 365L4200
Expédition estimée: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Thermo Scientific™ Gastight GC Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles are stainless steel
  • Plungers are PTFE
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 22s G
Length (Metric) 50 mm
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Quantity 1/Pk.
Volume (Metric) 2.5 mL
Length (Metric) Needle 56 mm
Needle Point Style Side Hole
For Use With (Equipment) CTC GC Instruments
Connection Type Fixed