Learn More
Invitrogen™ GeneBLAzer™ In Vivo Detection Kit
Product Code. 10432862
Change view
Click to view available options
Quantity:
375 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10432862 375 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10432862 Supplier Invitrogen™ Supplier No. 12578134
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
GeneBLAzer™ In Vivo Detection Kit
The GeneBLAzer™ In Vivo Detection Kit measures β-lactamase reporter activity in live mammalian cells, using the β-lactamase substrate CCF2-AM. The GeneBLAzer™ Detection System provides the following advantages:
  • Suitable for use as a sensitive reporter of gene expression in living mammalian cells using fluorescence microscopy.
  • Provides a ratiometric readout to minimize differences due to variability in cell number, substrate concentration, fluorescence intensity, and emission sensitivity
  • Uses a non-toxic substrate, allowing continued cell culturing after quantitative analysis.
  • Compatible with a wide variety of applications, including microplate-based transcriptional assays and flow cytometry.
  • Provides a flexible and simple assay development platform for gene expression in mammalian cells.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Content And Storage In vivo detection kits contain 50 μg of CCF2-AM substrate (enough for 375 96-well reactions). Substrate should be stored at -20°C, dessicated, and protected from light. All reagents are guaranteed stable 6 months when properly stored.
    Detection Method Fluorescence
    Product Type Detection Kit
    Quantity 375 Reactions
    Format Kit
    Key Functions Reporter Assays
    Reporter Gene BLA (Beta-Lactamase)
    Substrate Beta-Lactamase Substrate
    Product Line GeneBLAzer
    For Research Use Only. Not for use in diagnostic procedures.