Learn More
Aalborg™ TIO-GFM Mounting Kit
Product Code. 13594500
Change view
Click to view available options
Type:
GFM Mass Flow Meter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
13594500 GFM Mass Flow Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13594500 Supplier Aalborg™ Supplier No. KITTMDD
  • ONLINE EXCLUSIVE PRICE
    68.39€
    49.98€ / 1 pièces
    Économisez 18.41€
    26% de réduction
 Restricted Item
Expédition estimée: 27-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Designed especially for the laboratory these kits offer a diverse economical way of acquiring flow measurement capabilities. Aalborg™ TIO-GFM Mounting Kit allows for easy changing of flow tubes in the field.
  • Kits are shipped in convenient molded plastic carrying cases.
  • Flow tubes and floats are interchangeable in frames supplied facilitating overlapping flow ranges.
  • For non-corrosive fluids use the Aluminum Kit.
  • For corrosives consider the Stainless Steel Kit.
  • For corrosive applications at lower pressure or for high purity fluid service specify the PTFE Kit.
  • Supplied with glass floats installed.
Specifications
Type GFM Mass Flow Meter
For Use With (Application) Measures flow rate