Learn More
Gilac™ Gilactiv™ Patient Drinking Cup, Blue
Product Code. 17968431
Change view
Click to view available options
Capacity (Metric):
0.25 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17968431 0.25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17968431 Supplier Gilac™ Supplier No. G128490
  • ONLINE EXCLUSIVE PRICE
    6.09€
    4.05€ / 1 pièces
    Économisez 2.04€
    33% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Gilactiv™ Patient Drinking Cup is treated with the exclusive Sanitized™ technology, which protects the patient drinking cup's surface against viruses and bacteria.
  • Gilactiv™ patient drinking cup with graduations (in mL) treated with the exclusive Sanitized™ technology, which protects the patient drinking cup's surface against viruses and bacteria.
  • Proven antibacterial and antiviral effectiveness on the patient drinking cup's surface (empty or full) for a broad spectrum of germs, including HCoV-229E human coronavirus.
  • This article contains a biocidal product with the active substance: Silver Phosphate Glass.
  • Tests conducted by an EU-approved French independent laboratory showed that 98.6% of the virus was wiped out after a two-hour period.
  • Choose Gilactiv™ enhanced protection.
    • Test results available on request.
  • Integrated lid and drinking straw.
  • Allows drinking without spilling content.
Specifications
Material HDPE
Capacity (Metric) 0.25 L
Capacity (English) 8.5 oz.
Product Type Tumbler Cup