Learn More
Brady™ Miniature Circuit Breaker Lockouts - Pin-Out Wide
Product Code. 15613517 Shop All Brady Products
Change view
Click to view available options
Color:
Red/Black
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15613517 Red/Black Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613517 Supplier Brady™ Supplier No. 090851
  • ONLINE EXCLUSIVE PRICE
    125.00€
    120.30€ / 6 pièces
    Économisez 4.70€
    3% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
A safe and effective method for locking out European Circuit Breakers.
  • Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a thumb wheel for quick application
Specifications
Product Type Circuit Breaker Lockout
Alternate Part Number ELEC. CIRCUIT BREAKER L/O EURO POW PK6
Color Red/Black
Material Fiberglass-reinforced Plastic
Certifications/Compliance UL 94 HB
Description Miniature Circuit Breaker Lockouts - Pin-Out Wide
Material (Shackle) Fiberglass-reinforced Plastic
Quantity 6 Pk.