Learn More
DWK Life Sciences KIMBLE™ Glass-filled Nylon Connectors with PTFE Seals
Product Code. 11882933 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
13-425 to 15-425
13-425 to 20-400
15-425 to 20-400
18-400 to 20-400
20-400 to 20-400
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
11882933 15-425 to 20-400 Each
11892933 18-400 to 20-400 Each
17751088 13-425 to 20-400 Each
15389796 13-425 to 15-425 Each
11802943 20-400 to 20-400 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11882933 Supplier DWK Life Sciences Supplier No. 7472051520
  • ONLINE EXCLUSIVE PRICE
    22.22€
    20.03€ / 1 pièces
    Économisez 2.19€
    9% de réduction
 Restricted Item
En stock
Add to basket
Edge
Add to basket
KIMBLE™ Glass-filled Nylon Connectors with PTFE Seals are threaded connecting adapters for coupling two items with GPI-threaded finishes. Made from blue glass-filled nylon with PTFE seals and FKM O-rings, these connectors are rated for use up to 150°C.
  • Threaded connecting adapters for coupling two GPI-threaded components
  • Blue glass-filled nylon construction
  • PTFE seals and FKM O-rings for reliable sealing
  • Rated for use up to 150°C
  • Available in a range of GPI thread size combinations
Specifications
For Use With (Application) To connect standard rotary evaporator traps to vials
Includes PTFE seals
Thread Size 15-425 to 20-400
Material Nylon
Product Type Connector