Learn More
Hach Lange™ Cellules d’échantillon de verre
Easy to use and cost effective replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters are made of glass and contain six sample cell vials with caps.
88.50€
| Code produit | Marque | dimForUseWithEquipment | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | dimForUseWithEquipment | Prix | Quantité et disponibilité | |||||
|
10020362
|
Hach Lange™
2434706 |
105.00€ 88.50€ / 6 pièces Économisez 16.50€ 15% de réduction |
Expédition estimée: 22-04-2026
Connectez-vous pour voir le stock disponible |
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||||
Description
Easy to use and cost effective replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters are made of glass and contain six sample cell vials with caps.
- For use with Hach Ratio 18900, 2100N and 2100AN Laboratory Turbidmeters
- Replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters
- For use with Hach Ratio 18900, 2100N and 2100AN laboratory turbidmeters
- Includes six sample cell vials with caps
- Made of glass material