Learn More
Hecht Karl™ Glass Staining Dish
Product Code. 10682951
Change view
Click to view available options
Dimensions (L x W x H):
105 x 85 x 70 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
10682951 105 x 85 x 70 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10682951 Supplier Hecht Karl™ Supplier No. 42480010
  • ONLINE EXCLUSIVE PRICE
    32.37€
    26.66€ / 1 pièces
    Économisez 5.71€
    17% de réduction
Expédition estimée: 30-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type Staining Dish
Dimensions (L x W x H) 105 x 85 x 70 mm
Height (Metric) 70 mm
Length (Metric) 105 mm
Material Glass
Quantity 1 Ea.
Width (Metric) 85 mm