Learn More
Labconco Glass Trap
Product Code. 18891916 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material
18891916 Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18891916 Supplier Labconco Supplier No. L7397605
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Protects CentriVap Cold Trap reservoir and lid from corrosives
  • Included with CentriVap Acid Systems
  • Not compatible with pre-2005 Cold Traps or Mobile Systems.
  • Shipping weight 4.0 lb. (1.8 kg)
Specifications
Material Glass
Type Glass Trap
For Use With (Equipment) CentriVap Cold Trap reservoir and lid