Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10
Beskrivelse
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis.
- Made from optically clear plastic
- Glasstic Slide features 10 individual fully segregated specimen wells
- Capillary action reduces the likelihood of pipetting errors
- Suspends sediments homogeneously throughout the specimen
- Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
- Reproducible fluid distribution throughout each chamber
- Chamber walls prevent specimen leakage and cross-contamination
- Correlate well with results obtained by using a hemocytometer: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Tekniske data
Tekniske data
| Type | Slide 10 |
| For Use With (Application) | Microscopy |
| Gridded | No |
| Material | Plastic with the optical qualities of glass |
| No. of Wells | 10 |
| Quantity | 100 Pack |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?