Learn More
Globe Scientific Flanged Plug Caps For 16mm Tubes
Product Code. 31922745 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Black
Blue
Green
Lavender
Orange
Red
White
Unit Size:
Pack of 10000
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31922745 Black Pack of 10000
31923034 Green Pack of 10000
31922571 Lavender Pack of 10000
31922027 Blue Pack of 10000
31922551 White Pack of 10000
31921871 Red Pack of 10000
31923006 Orange Pack of 10000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31922745 Supplier Globe Scientific Supplier No. 116152K
Expédition estimée: 11-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes
  • Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
  • Available in seven colors for sample identification purposes
Specifications
Includes 2 flexible flanges
Product Type Flange Plug Cap
Material Polyethylene
Color Black
Size (English) 0.62 in.
Size (Metric) 16 mm
For Use With (Equipment) 16 mm Tubes