Learn More
Glycerol Triformate, TRC
Product Code. 30135384
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135384 5 g 5g
30010775 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135384 Supplier Toronto Research Chemicals Supplier No. TRCG6015455G
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32765-69-8
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
Synonym Triformin, 1,2,3-Propanetriol, 1,2,3-Triformate, 1,2,3-Propanetriol, Triformate
IUPAC Name 2,3-diformyloxypropyl formate
SMILES O=COCC(COC=O)OC=O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H8O6/c7-3-10-1-6(12-5-9)2-11-4-8/h3-6H,1-2H2
Formula Weight 176.0321
Percent Purity >95%
Chemical Name or Material Glycerol Triformate
Grade GC