missing translation for 'onlineSavingsMsg'
Learn More

Glycyl diphenylborinate, 95%, Thermo Scientific™

Product Code. 15421688
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421688 1 g 1g
15431688 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421688 Supplier Thermo Scientific Alfa Aesar Supplier No. H55443.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14335-29-6
Molecular Formula C14H14BNO2
Molecular Weight (g/mol) 239.081
MDL Number MFCD05863277
InChI Key IVHLNYVPKWICDY-UHFFFAOYSA-N
Synonym 2-diphenylboryl oxy-2-oxoethylamine, glycyl diphenylborinate, glycine diphenylboryl ester, diphenylboramethyl 2-aminoacetate, 2-diphenylboryl oxy-2-oxoethanamine
PubChem CID 11840443
IUPAC Name diphenylboranyl 2-aminoacetate
SMILES B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)CN

Specifications

Quantity 1 g
Formula Weight 239.08
Percent Purity 95%
Chemical Name or Material Glycyl diphenylborinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.