Learn More
Gosselin™ 100x15mm Bi-Plate Petri Dish, Double Bagged with Traceability Labels, Optimized for automation
BIOPHARMA
Product Code. 15448784
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 825
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
15448784 Sterile Pack of 825
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448784 Supplier Gosselin™ Supplier No. BI93D102
  • ONLINE EXCLUSIVE PRICE
    169.59€
    161.24€ / 825 pièces
    Économisez 8.35€
    4% de réduction
Expédition estimée: 13-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Optimized for automation and double-bagged for security
Sterile product features a double outer bag with traceability.
  • Three vents
  • Crystal polystyrene
  • Optimal flatness
  • Manufactured in class 100
  • Frosted area
  • Easy-opening sleeves
TRUSTED_SUSTAINABILITY
Specifications
Description Petri dish
Disposable Single-use
Material Polystyrene
Shape Round
Sterility Sterile
Well Count 2
Packaging Easy-open sleeves, double-bagged with Traceability Labels
Vent 3
Height (Metric) 14.2 mm
Diameter (Metric) 90 mm