Learn More
WTW™ Graphite 2-Electrode Conductivity Cell
Product Code. 10540724
Change view
Click to view available options
Material:
Graphite
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10540724 Graphite Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10540724 Supplier WTW™ Supplier No. 301995
  • ONLINE EXCLUSIVE PRICE
    585.95€
    498.00€ / 1 pièces
    Économisez 87.95€
    15% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
The universally usable conductivity measuring cells by WTW set standard in the conductivity measurement. Their 4 electrode technology makes them insensitive to pollution, uninfluenced by polarization effects and therefore precise and reliable in any conditions.
TRUSTED_SUSTAINABILITY
Specifications
Description 2-electrode graphite conductivity cell
For Use With (Equipment) Standard KLE 325 Electrode
For Use With (Application) Eliminate errors caused by polarization effects, and from dirty samples
Includes Built-in temperature sensor and 1.5m cable
Material Graphite
Temperature (Metric) Operating 0°C to 80°C
Length (Metric) Cable 1.5 m