Få mere at vide
Checkers Grip Guard™ 3 Channel Lightweight Cable Protector
Artikelnummer. 17772418
Skift visning
Klik for at se tilgængelige muligheder
Color:
Yellow, Gray
Pakningsstørrelse:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Color Pakningsstørrelse
17772418 Yellow, Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17772418 Leverandør Checkers Leverandørnr. PGGS0001YG
  • ONLINE EXCLUSIVE PRICE
    364.55€
    296.13€ / 1 pièces
    Économisez 68.42€
    18% de réduction
Expédition estimée: 26-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
A lightweight, yet durable cable protector designed for pedestrian and light vehicle traffic.
  • Grip Guard™ series 3-channel cable protectors feature our 'Gripper Connectors' with a new lightweight design
  • At a weight of only 104 kg, Grip Guard™ is up to 33% lighter than other leading polyurethane 3-channel cable protectors, and up to 50% lighter than other rubber 3-channel cable protectors on the market
  • The Gripper Connector is ergonomically designed to fit your hand, which allows for easy handling and transport
  • This connector also locks multiple cable protectors tightly together to provide a safe crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines
  • When compared to other rubber manufactured cable protectors, Grip Guard™ is up to 50% lighter without compromising the required strength and durability
Tekniske data
Length (English) 36 in.
Length (Metric) 91 cm
Width (Metric) 46 cm
Color Yellow, Gray
Material Polyurethane
Type Cable Protector
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.