Learn More
Checkers Guard Dog™ ADA 5 Channel Cable Protector
Product Code. 17762408
Change view
Click to view available options
Color:
Black, Orange
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17762408 Black, Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17762408 Supplier Checkers Supplier No. PGDS0006OB
  • ONLINE EXCLUSIVE PRICE
    376.85€
    306.75€ / 1 pièces
    Économisez 70.10€
    18% de réduction
Expédition estimée: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Guard Dog series 5-channel cable protector with a hinged lid provides a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines from damage.
  • Guard Dog series 5-channel cable protector with hinged lid provides a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines from damage
  • Modular interlocking design is ideal for use in amusement parks, entertainment venues, sporting events, and most public, commercial, industrial, construction, and utility applications
Specifications
For Use With (Application) Safer Crossing for Vehicle and Pedestrian Traffic
Includes Hinged Lid
Length (English) 151 ft.
Length (Metric) 46 m
Width (Metric) 127 cm
Color Black, Orange
Type Cable Protector